Triethylene glycol monobenzyl ether ≥95.0%

Supplier: TCI America

T1996-5G T1996-25G
TCT1996-5GEA 279.34 CAD
TCT1996-5G TCT1996-25G
Triethylene glycol monobenzyl ether ≥95.0%
Triethylene glycol monobenzyl ether

CAS Number: 55489-58-2
MDL Number: MFCD02093556
Molecular Formula: C13H20O4
Molecular Weight: 240.30
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol

Formula: C₁₃H₂₀O₄
Density: 1.09 (20 °C)
MDL Number: MFCD02093556
CAS Number: 55489-58-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed9dba8d6ecd5e189d54da699bc8f3ac8d07426f&vid=ed9dba8d6ecd5e189d54da699bc8f3ac8d07426f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR