2,3,4,6-Tetra-O-benzoyl-ɑ-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate 96,0

Supplier: TCI America

T1991-200MG T1991-1G
TCT1991-200MGEA 772.62 CAD
TCT1991-200MG TCT1991-1G
2,3,4,6-Tetra-O-benzoyl-ɑ-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate 96,0
2,3,4,6-Tetra-O-benzoyl-alpha-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate

CAS Number: 428816-48-2
MDL Number: MFCD06797174
Molecular Formula: C50H37F6NO10S
Molecular Weight: 957.89
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Color: White
Storage Temperature: <0°C

Formula: C₅₀H₃₇F₆NO₁₀S
MW: 957.89 g/mol
MDL Number: MFCD06797174
CAS Number: 428816-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edefeb491de422ec7783cb440247429d55481dd6&vid=edefeb491de422ec7783cb440247429d55481dd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR