4'-Methoxy-2,2,2-trifluoroacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 2,2,2-Trifluoro-1-(4-methoxyphenyl)ethanone

T1986-25G T1986-5G
TCT1986-25GEA 808.33 CAD
TCT1986-25G TCT1986-5G
4'-Methoxy-2,2,2-trifluoroacetophenone ≥98.0%
4'-Methoxy-2,2,2-trifluoroacetophenone

CAS Number: 711-38-6
MDL Number: MFCD00018061
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.32


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4'-Methoxy-2,2,2-trifluoroacetophenone, 4-(Trifluoroacetyl)anisole

Formula: C₉H₇F₃O₂
MW: 204.1500 g/mol
Density: 1.32 (20 °C)
MDL Number: MFCD00018061
CAS Number: 711-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14e0497018d81dfc0edb31de0a1785f112479bd7&vid=14e0497018d81dfc0edb31de0a1785f112479bd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR