3-Ethyl-(-)-thiazolidine-3,4-dicarboxylate ≥98.0%

Supplier: TCI America

Synonyms: (-)-3-Carbethoxythiazolidine-4-carboxylic Acid, (-)-3-Ethoxycarbonylthiazolidine-4-carboxylic Acid, (-)-Thiazolidine-3,4-dicarboxylic Acid 3-Ethyl Ester

T1974-25G T1974-5G
TCT1974-25GEA 102.7 CAD
TCT1974-25G TCT1974-5G
3-Ethyl-(-)-thiazolidine-3,4-dicarboxylate ≥98.0%
3-Ethyl-(-)-thiazolidine-3,4-dicarboxylate

CAS Number: 127657-29-8
MDL Number: MFCD04117921
Molecular Formula: C7H11NO4S
Molecular Weight: 205.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 98
Specific rotation [a]20/D: -120 deg (C=1, H2O)


Other Product Names / Synonyms: (-)-3-Carbethoxythiazolidine-4-carboxylic Acid, (-)-3-Ethoxycarbonylthiazolidine-4-carboxylic Acid, (-)-Thiazolidine-3,4-dicarboxylic Acid 3-Ethyl Ester

Formula: C₇H₁₁NO₄S
MW: 205.23 g/mol
Melting Pt: 98 °C
MDL Number: MFCD04117921
CAS Number: 127657-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5fa7805d9ab949382a722e3c4033e831cc08d1d&vid=c5fa7805d9ab949382a722e3c4033e831cc08d1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR