1,4,8,11-Tetrathiacyclotetradecane ≥98.0%

Supplier: TCI America

T1959-1G T1959-200MG
TCT1959-1GEA 924.3 CAD
TCT1959-1G TCT1959-200MG
1,4,8,11-Tetrathiacyclotetradecane ≥98.0%
1,4,8,11-Tetrathiacyclotetradecane

CAS Number: 24194-61-4
MDL Number: MFCD00010048
Molecular Formula: C10H20S4
Molecular Weight: 268.51
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 123

Formula: C₁₀H₂₀S₄
MW: 268.51 g/mol
Melting Pt: 123 °C
MDL Number: MFCD00010048
CAS Number: 24194-61-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0037d18cf6d104acede6993afedaf01ecf4a49e&vid=b0037d18cf6d104acede6993afedaf01ecf4a49e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR