Tri-o-tolylbismuth dichloride ≥97.0%

Supplier: TCI America

Synonyms: Dichloro(tri-o-tolyl)bismuth, Tris(2-methylphenyl)bismuth Dichloride

T1957-1G T1957-5G
TCT1957-1GEA 307.47 CAD
TCT1957-1G TCT1957-5G
Tri-o-tolylbismuth dichloride ≥97.0%
Tri-o-tolylbismuth dichloride

CAS Number: 6729-60-8
MDL Number: MFCD00092459
Molecular Formula: C21H21BiCl2
Molecular Weight: 553.28
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Hypervalent Bismuth Compounds, Oxidation, Oxidation
Other Product Names / Synonyms: Dichloro(tri-o-tolyl)bismuth, Tris(2-methylphenyl)bismuth Dichloride

Formula: C₂₁H₂₁BiCl₂
MDL Number: MFCD00092459
CAS Number: 6729-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3e1a647bc4c5aa35aed660f7119fbfc5b276fc8&vid=c3e1a647bc4c5aa35aed660f7119fbfc5b276fc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR