3-Hydroxyphenylboronic acid pinacol ester ≥98.0%
Supplier: TCI America
Synonyms:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
CAS Number: 214360-76-6
MDL Number: MFCD02093755
Molecular Formula: C12H17BO3
Molecular Weight: 220.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 99
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(3-Hydroxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hydroxyphenylboronic Acid Pinacol Ester
Formula:
C₁₂H₁₇BO₃ MW: 220.08 g/mol Melting Pt: 99 °C |
MDL Number:
MFCD02093755 CAS Number: 214360-76-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...