2-Hydroxyphenylboronic acid pinacol ester ≥97.0%

Supplier: TCI America

T1952-1G T1952-5G
TCT1952-1GEA 132.7 CAD
TCT1952-1G TCT1952-5G
2-Hydroxyphenylboronic acid pinacol ester ≥97.0%
2-Hydroxyphenylboronic acid pinacol ester

CAS Number: 269409-97-4
MDL Number: MFCD02093754
Molecular Formula: C12H17BO3
Molecular Weight: 220.08
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 94
Specific Gravity (20/20): 1.06


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(2-Hydroxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Hydroxyphenylboronic Acid Pinacol Ester

Formula: C₁₂H₁₇BO₃
Boiling Pt: 94 °C
Density: 1.06 (20 °C)
MDL Number: MFCD02093754
CAS Number: 269409-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfe10a1001fe746867222d9b831bed35263f949a&vid=bfe10a1001fe746867222d9b831bed35263f949a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR