2,4,6-Tri(4-pyridyl)-1,3,5-triazine ≥97.0% (by total nitrogen basis), purified by sublimation

Supplier: TCI America

T1937-5G T1937-1G
TCT1937-5GEA 615.55 CAD
TCT1937-5G TCT1937-1G
2,4,6-Tri(4-pyridyl)-1,3,5-triazine ≥97.0% (by total nitrogen basis), purified by sublimation
2,4,6-Tri(4-pyridyl)-1,3,5-triazine

CAS Number: 42333-78-8
MDL Number: MFCD00129048
Molecular Formula: C18H12N6
Molecular Weight: 312.34
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Color: White

Formula: C₁₈H₁₂N₆
MW: 312.34 g/mol
MDL Number: MFCD00129048
CAS Number: 42333-78-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1c40eabea6ce989c936d0a28dbf47593721c566&vid=1fd3b7efe5c9397a8b395b46801f1406acea1e95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR