Sodium 8-anilinonaphthalene-1-sulfonate ≥97.0%

Supplier: TCI America

Synonyms: ANS-Na, 8-Anilino-1-naphthalenesulphonic acid sodium salt, Sodium 8-Anilino-1-naphthalenesulphonate, 8-Anilino-1-naphthalenesulfonic acid sodium salt, Sodium 8-Anilino-1-naphthalenesulfonate

A5352-1G A5352-5G
TCA5352-001GEA 66.36 CAD
TCA5352-001G TCA5352-5G
Sodium 8-anilinonaphthalene-1-sulfonate ≥97.0%
Sodium 8-anilinonaphthalene-1-sulphonate

[Hydrophobic fluorescent probe]
CAS Number: 1445-19-8
MDL Number: MFCD00054279
Molecular Formula: C16H13NO3S
Molecular Weight: 321.33
Purity/Analysis Method: >97.0% (N)
Form: Crystal


Product Categories: Bioscience, Fluorescent Probes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 8-Anilino-1-naphthalenesulfonic Acid Sodium Salt, Sodium 8-Anilino-1-naphthalenesulfonate

Formula: C₁₆H₁₂NNaO₃S
MDL Number: MFCD00054279
CAS Number: 1445-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87be8c122ab9522b7e7526df4ea372827db811f1&vid=bf551a3e57a8a0a0f1cb9a2a780c646b34cf8d4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR