2,4,6-Tris(3,4,5-trifluorophenyl)boroxin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tris(3,4,5-trifluorophenyl)cycloboroxane, 3,4,5-Trifluorophenylboronic Anhydride

T1929-1G T1929-5G
TCT1929-1GEA 255.96 CAD
TCT1929-1G TCT1929-5G
2,4,6-Tris(3,4,5-trifluorophenyl)boroxin ≥98.0% (by titrimetric analysis)
2,4,6-Tris(3,4,5-trifluorophenyl)boroxin

CAS Number: 223440-94-6
MDL Number: MFCD03844809
Molecular Formula: C18H6B3F9O3
Molecular Weight: 473.66
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 3,4,5-Trifluorophenylboronic Anhydride, Tris(3,4,5-trifluorophenyl)cycloboroxane

Formula: C₁₈H₆B₃F₉O₃
MW: 473.66 g/mol
MDL Number: MFCD03844809
CAS Number: 223440-94-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3965399543ef55abae5c162ed14ad26151a52b94&vid=3965399543ef55abae5c162ed14ad26151a52b94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR