1-[3-(Trimethoxysilyl)propyl]urea ≥94.0%

Supplier: TCI America

Synonyms: (3-Ureidopropyl)trimethoxysilane

T1915-250G T1915-25G
TCT1915-250GEA 446.46 CAD
TCT1915-250G TCT1915-25G
1-[3-(Trimethoxysilyl)propyl]urea ≥94.0%
1-[3-(Trimethoxysilyl)propyl]urea

CAS Number: 23843-64-3
MDL Number: MFCD00216654
Molecular Formula: C7H18N2O4Si
Molecular Weight: 222.32
Purity/Analysis Method: >94.0% (N)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 98
Specific Gravity (20/20): 1.15


Product Categories: Synthetic Organic Chemistry, Ureas (Multi-component Reaction), Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: (3-Ureidopropyl)trimethoxysilane

Formula: C₇H₁₈N₂O₄Si
MW: 222.32 g/mol
Density: 1.15 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00216654
CAS Number: 23843-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b969bdb46f09d2cf0a65dccfb490ee24dd3ed5c6&vid=b969bdb46f09d2cf0a65dccfb490ee24dd3ed5c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR