6-Bromo-2-naphthyl trifluoromethanesulfonate ≥95.0%

Supplier: TCI America

Synonyms: 6-Bromo-2-naphthyl Triflate, Trifluoromethanesulphonic acid 6-Bromo-2-naphthyl Ester, Trifluoromethanesulfonic acid 6-Bromo-2-naphthyl Ester, 6-Bromonaphthalen-2-yl trifluoromethanesulfonate, 6-Bromonaphthalen-2-yl trifluoromethanesulphonate

T1894-5G T1894-25G
TCT1894-5GEA 221.52 CAD
TCT1894-5G TCT1894-25G
6-Bromo-2-naphthyl trifluoromethanesulfonate ≥95.0%
6-Bromo-2-naphthyl trifluoromethanesulfonate

CAS Number: 151600-02-1
MDL Number: MFCD03093642
Molecular Formula: C11H6BrF3O3S
Molecular Weight: 355.13
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 54


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 6-Bromo-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 6-Bromo-2-naphthyl Ester

Formula: C₁₁H₆BrF₃O₃S
Melting Pt: 54 °C
MDL Number: MFCD03093642
CAS Number: 151600-02-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4b060ef3cf692bc7e71054e3da7c630f3600778&vid=c4b060ef3cf692bc7e71054e3da7c630f3600778&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR