Ammonium 8-anilinonaphthalene-1-sulfonate ≥95.0%

Supplier: TCI America

Synonyms: Ammonium 8-(phenylamino)naphthalene-1-sulphonate, Ammonium 8-(phenylamino)naphthalene-1-sulfonate, 8-Anilino-1-naphthalenesulphonic acid Ammonium salt, 8-Anilinonaphthalene-1-sulfonic acid ammonium salt, 8-Anilino-1-naphthalenesulfonic acid Ammonium salt, Ammonium 8-Anilino-1-naphthalenesulphonate, Ammonium 8-Anilino-1-naphthalenesulfonate, 8-Anilinonaphthalene-1-sulphonic acid ammonium salt, ANS-NH4

A5351-1G A5351-10G
TCA5351-001GEA 45.82 CAD
TCA5351-001G TCA5351-010G
Ammonium 8-anilinonaphthalene-1-sulfonate ≥95.0%
Ammonium 8-anilinonaphthalene-1-sulphonate

[Hydrophobic fluorescent probe]
CAS Number: 28836-03-5
MDL Number: MFCD00012560
Molecular Formula: C16H16N2O3S
Molecular Weight: 316.38
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 237
Lambda max.: 375 nm


Product Categories: Bioscience, Fluorescent Probes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Ammonium 8-Anilino-1-naphthalenesulfonate, 8-Anilino-1-naphthalenesulfonic Acid Ammonium Salt

Formula: C₁₆H₁₆N₂O₃S
MW: 316.37 g/mol
Melting Pt: 237 °C
MDL Number: MFCD00012560
CAS Number: 28836-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87da2699dc6228db311b038bd54fde416bc7da25&vid=541a5d4b2b8597c951706e7410f946768f022937&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR