3,4,5-Tris(benzyloxy)benzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T1869-25G T1869-5G
TCT1869-25GEA 955.9 CAD
TCT1869-25G TCT1869-5G
3,4,5-Tris(benzyloxy)benzoic acid ≥98.0% (by HPLC, titration analysis)
3,4,5-Tris(benzyloxy)benzoic acid

CAS Number: 1486-48-2
MDL Number: MFCD00825832
Molecular Formula: C28H24O5
Molecular Weight: 440.50
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 196


Other Product Names / Synonyms: 3,4,5-Tribenzyloxybenzoic Acid

Formula: C₂₈H₂₄O₅
Melting Pt: 196 °C
MDL Number: MFCD00825832
CAS Number: 1486-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4139e99d93d1d9f91ef50775d58a67c969f7b64&vid=b4139e99d93d1d9f91ef50775d58a67c969f7b64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR