(1R)-(-)-Thiocamphor ≥97.0%

Supplier: TCI America

T1863-5G T1863-1G
TCT1863-005GEA 534.04 CAD
TCT1863-005G 76220-510
(1R)-(-)-Thiocamphor ≥97.0%
(1R)-(-)-Thiocamphor

CAS Number: 53402-10-1
MDL Number: MFCD00074855
Molecular Formula: C10H16S
Molecular Weight: 168.30
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: Yellow
Melting point (°C): 137
Specific rotation [a]20/D: -21 deg (C=3, EtOAc)

Formula: C₁₀H₁₆S
MW: 168.30 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00074855
CAS Number: 53402-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d846ac2e42f981bf17d457c57cc8883ae824cab7&vid=d846ac2e42f981bf17d457c57cc8883ae824cab7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR