Tri-O-benzyl-D-glucal ≥95.0% (by HPLC)

Supplier: TCI America

T1859-5G T1859-1G
TCT1859-005GEA 845.3 CAD
TCT1859-005G TCT1859-001G
Tri-O-benzyl-D-glucal ≥95.0% (by HPLC)
Tri-O-benzyl-D-glucal

CAS Number: 55628-54-1
MDL Number: MFCD00061640
Molecular Formula: C27H28O4
Molecular Weight: 416.52
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 57
Specific rotation [a]20/D: -2 deg (C=2.5, CH3CN)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Glucose, Glycals
Other Product Names / Synonyms: 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tribenzyl Ether

Formula: C₂₇H₂₈O₄
Melting Pt: 57 °C
MDL Number: MFCD00061640
CAS Number: 55628-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86b9027f2e2c384d365c261ec5b7f19e0a0a3369&vid=86b9027f2e2c384d365c261ec5b7f19e0a0a3369&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR