5-Aminofluorescein (isomer I) ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one (isomer I), 5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-Aminofluorescein, Fluoresceinamine, isomer I, 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, Fluoresceinamine (isomer I)
CAS Number: 3326-34-9
MDL Number: MFCD00005052
Molecular Formula: C20H13NO5
Molecular Weight: 347.33
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Product Categories: Bioscience, Fluorometry & Fluorescent Imaging, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, Fluoresceinamine
Formula:
C₂₀H₁₃NO₅ MW: 347.33 g/mol |
MDL Number:
MFCD00005052 CAS Number: 3326-34-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...