(+)-N,N,N',N'-Tetramethyl-L-tartardiamide ≥98.0%

Supplier: TCI America

T1839-5G T1839-25G
TCT1839-005GEA 104.28 CAD
TCT1839-005G TCT1839-025G
(+)-N,N,N',N'-Tetramethyl-L-tartardiamide ≥98.0%
(+)-N,N,N',N'-Tetramethyl-L-tartardiamide

CAS Number: 26549-65-5
MDL Number: MFCD00025672
Molecular Formula: C8H16N2O4
Molecular Weight: 204.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 187
Specific rotation [a]20/D: 45 deg (C=1, EtOH)


Other Product Names / Synonyms: (R,R)-(+)-2,3-Dihydroxy-N,N,N',N'-tetramethylsuccinamide

Formula: C₈H₁₆N₂O₄
Melting Pt: 187 °C
MDL Number: MFCD00025672
CAS Number: 26549-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad2be9ed4a0fb82de2f15dc3f07d9812a4693bd7&vid=ad2be9ed4a0fb82de2f15dc3f07d9812a4693bd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR