Triphenylbismuth carbonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1835-1G T1835-5G
TCT1835-1GEA 185.18 CAD
TCT1835-1G TCT1835-5G
Triphenylbismuth carbonate ≥98.0% (by titrimetric analysis)
Triphenylbismuth carbonate

CAS Number: 47252-14-2
MDL Number: MFCD00209622
Molecular Formula: C19H15BiO3
Molecular Weight: 500.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Hypervalent Bismuth Compounds
Other Product Names / Synonyms: [Carbonato(2-)-O]triphenylbismuth

Formula: C₁₉H₁₅BiO₃
MDL Number: MFCD00209622
CAS Number: 47252-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d32253eff06bb0ffb138cea4c2ebe9eafe269e5&vid=1d32253eff06bb0ffb138cea4c2ebe9eafe269e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR