Dimethyl tetrahydropyran-4,4-dicarboxylate ≥98.0%

Supplier: TCI America

Synonyms: Tetrahydropyran-4,4-dicarboxylic Acid Dimethyl Ester, Dimethyl Tetrahydropyran-4,4-dicarboxylate

T1825-5G T1825-25G
TCT1825-005GEA 192.76 CAD
TCT1825-005G TCT1825-25G
Dimethyl tetrahydropyran-4,4-dicarboxylate ≥98.0%
Dimethyl tetrahydropyran-4,4-dicarboxylate

CAS Number: 149777-00-4
MDL Number: MFCD02093499
Molecular Formula: C9H14O5
Molecular Weight: 202.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.18


Other Product Names / Synonyms: Tetrahydropyran-4,4-dicarboxylic Acid Dimethyl Ester

Formula: C₉H₁₄O₅
MW: 202.21 g/mol
Density: 1.18 (20 °C)
MDL Number: MFCD02093499
CAS Number: 149777-00-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f844d78199a3db8c5e2461aae3e8b1c20d0384e6&vid=f844d78199a3db8c5e2461aae3e8b1c20d0384e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR