Tetracyclo[6.2.1.1(3,6).0(2,7)]dodec-4-ene ≥95.0%
Supplier: TCI America
T1817-5G
T1817-1G
T1817-25G
TCT1817-5GEA
378.46
CAD
TCT1817-5G
TCT1817-001G
TCT1817-25G
Tetracyclo[6.2.1.1(3,6).0(2,7)]dodec-4-ene ≥95.0%
Tetracyclo[6.2.1.1(3,6).0(2,7)]dodec-4-ene
CAS Number: 21635-90-5
MDL Number: MFCD00213460
Molecular Formula: C12H16
Molecular Weight: 160.26
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 93
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: 1,2,3,4,4a,5,8,8a-Octahydro-1,4:5,8-dimethanonaphthalene
Formula:
C₁₂H₁₆ Boiling Pt: 93 °C Density: 1.02 (20 °C) |
MDL Number:
MFCD00213460 CAS Number: 21635-90-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...