5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin ≥90.0% (by HPLC)
Supplier: TCI America
Synonyms:
5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)-21H,23H-porphine
CAS Number: 145764-54-1
MDL Number: MFCD02093497
Molecular Formula: C44H30N4O8
Molecular Weight: 742.74
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Color: Deep Blue
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)-21H,23H-porphine
Formula:
C₄₄H₃₀N₄O₈ MW: 742.74 g/mol |
MDL Number:
MFCD02093497 CAS Number: 145764-54-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...