N,N,N',N'-Tetraphenylbenzidine ≥98.0%

Supplier: TCI America

Synonyms: TPB, N,N,N,N-Tetraphenylbenzidine, 4,4'-Bis(diphenylamino)biphenyl

T1812-5G T1812-25G
TCT1812-005GEA 130.26 CAD
TCT1812-005G TCT1812-25G
N,N,N',N'-Tetraphenylbenzidine ≥98.0%
N,N,N',N'-Tetraphenylbenzidine

CAS Number: 15546-43-7
MDL Number: MFCD00228123
Molecular Formula: C36H28N2
Molecular Weight: 488.63
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 233
Lambda max.: 354 nm (CHCl3)


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: 4,4'-Bis(diphenylamino)biphenyl, TPB

Formula: C₃₆H₂₈N₂
MW: 488.63 g/mol
Melting Pt: 233 °C
MDL Number: MFCD00228123
CAS Number: 15546-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5da2e0812e0d49bfd10d8509f513ff9091c2dd1&vid=f5da2e0812e0d49bfd10d8509f513ff9091c2dd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR