3-(Trifluoroacetyl)indole

Supplier: TCI America

T1809-5G T1809-1G
TCT1809-005GEA 244.9 CAD
TCT1809-005G TCT1809-1G
3-(Trifluoroacetyl)indole
3-(Trifluoroacetyl)indole

CAS Number: 14618-45-2
MDL Number: MFCD00182114
Molecular Formula: C10H6F3NO
Molecular Weight: 213.16
Form: Crystal
Color: White

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
MDL Number: MFCD00182114
CAS Number: 14618-45-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05e257e1236434e8b2ec2b5396dcd2d30ad15591&vid=05e257e1236434e8b2ec2b5396dcd2d30ad15591&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR