Tetrabutylammonium perrhenate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1803-1G T1803-5G
TCT1803-1GEA 180.12 CAD
TCT1803-1G TCT1803-5G
Tetrabutylammonium perrhenate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium perrhenate

CAS Number: 16385-59-4
MDL Number: MFCD00012037
Molecular Formula: C16H36NO4Re
Molecular Weight: 492.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₁₆H₃₆NO₄Re
MDL Number: MFCD00012037
CAS Number: 16385-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20ca690258497c3ef1e642cf00951b8de06997bb&vid=20ca690258497c3ef1e642cf00951b8de06997bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR