(±)-1-Phenyl-2,2,2-trifluoroethanol ≥97.0% for synthesis

Supplier: TCI America

Synonyms: α-(Trifluoromethyl)benzyl alcohol, 2,2,2-Trifluoro-1-phenylethan-1-ol

T1792-5G T1792-25G
TCT1792-005GEA 265.44 CAD
TCT1792-005G TCT1792-25G
(±)-1-Phenyl-2,2,2-trifluoroethanol ≥97.0% for synthesis
(±)-1-Phenyl-2,2,2-trifluoroethanol

CAS Number: 340-05-6
MDL Number: MFCD00004498
Molecular Formula: C8H7F3O
Molecular Weight: 176.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 88
Flash Point (°C): 84
Specific Gravity (20/20): 1.30


Other Product Names / Synonyms: 1-Phenyl-2,2,2-trifluoroethanol

Formula: C₆H₅CH(OH)CF₃
Boiling Pt: 88 °C
Density: 1.30 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00004498
CAS Number: 340-05-6
EINECS: 206-429-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11532476ce588c2210ec2abcee027afba2d6998d&vid=11532476ce588c2210ec2abcee027afba2d6998d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR