Terephthalamic acid ≥95.0% (by HPLC)

Supplier: TCI America

T1768-5G T1768-25G
TCT1768-005GEA 440.82 CAD
TCT1768-005G TCT1768-25G
Terephthalamic acid ≥95.0% (by HPLC)
Terephthalamic acid

CAS Number: 6051-43-0
MDL Number: MFCD00025482
Molecular Formula: C8H7NO3
Molecular Weight: 165.15
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 248


Other Product Names / Synonyms: Terephthalamidic Acid

Formula: C₈H₇NO₃
Melting Pt: 248 °C
MDL Number: MFCD00025482
CAS Number: 6051-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf41225ee6f8c9b0bb34e39cd08180e14ec4f82b&vid=cf41225ee6f8c9b0bb34e39cd08180e14ec4f82b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR