Triphenylborane triphenylphosphine complex ≥97.0%

Supplier: TCI America

T1757-5G T1757-25G
TCT1757-5GEA 96.38 CAD
TCT1757-5G TCT1757-025G
Triphenylborane triphenylphosphine complex ≥97.0%
Triphenylborane triphenylphosphine complex

CAS Number: 3053-68-7
MDL Number: MFCD01631317
Molecular Formula: C36H30BP
Molecular Weight: 504.42
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White

Formula: C₃₆H₃₀BP
MW: 504.42 g/mol
MDL Number: MFCD01631317
CAS Number: 3053-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd22d2f1923a2781ab80a06fce389a1762bebfcb&vid=bd22d2f1923a2781ab80a06fce389a1762bebfcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR