Tetraethylammonium trifluoromethanesulfonate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
T1745-25G
T1745-5G
TCT1745-025GEA
496.12
CAD
TCT1745-025G
TCT1745-005G
Tetraethylammonium trifluoromethanesulfonate ≥99.0% (by titrimetric analysis)
Tetraethylammonium trifluoromethanesulphonate
CAS Number: 35895-69-3
MDL Number: MFCD00042586
Molecular Formula: C9H20F3NO3S
Molecular Weight: 279.32
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 160
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Tetraethylammonium Triflate
Formula:
C₉H₂₀F₃NO₃S Melting Pt: 160 °C |
MDL Number:
MFCD00042586 CAS Number: 35895-69-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...