N-(4-Aminobutyl)-N-ethylisoluminol ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 6-[N-(4-Aminobutyl)-N-ethylamino]-2,3-dihydro-1,4-phthalazinedione, ABEI

A5304-1G A5304-100MG
TCA5304-1GEA 1899.16 CAD
TCA5304-1G TCA5304-100MG
N-(4-Aminobutyl)-N-ethylisoluminol ≥97.0% (by titrimetric analysis)
N-(4-Aminobutyl)-N-ethylisoluminol

[Chemiluminescence Reagent]
CAS Number: 66612-29-1
MDL Number: MFCD00010559
Molecular Formula: C14H20N4O2
Molecular Weight: 276.34
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Product Categories: Analytical Science, Luminols (Chemiluminescence Reagents), Chemiluminescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: ABEI, 6-[N-(4-Aminobutyl)-N-ethylamino]-2,3-dihydro-1,4-phthalazinedione

Formula: C₁₄H₂₀N₄O₂
MW: 276.34 g/mol
MDL Number: MFCD00010559
CAS Number: 66612-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5337808ef73b033ac7ce9386d98c060d43fded9f&vid=5337808ef73b033ac7ce9386d98c060d43fded9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR