Dimethyl tetradecanedioate ≥96.0%
Supplier: TCI America
T1726-5G
T1726-25G
TCT1726-005GEA
178.54
CAD
TCT1726-005G
TCT1726-025G
Dimethyl tetradecanedioate ≥96.0%
Dimethyl tetradecanedioate
CAS Number: 5024-21-5
MDL Number: MFCD00673459
Molecular Formula: C16H30O4
Molecular Weight: 286.41
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 196
Melting point (°C): 43
Other Product Names / Synonyms: Tetradecanedioic Acid Dimethyl Ester
Formula:
C₁₆H₃₀O₄ Boiling Pt: 196 °C Melting Pt: 43 °C |
MDL Number:
MFCD00673459 CAS Number: 5024-21-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...