Tris(4-iodophenyl)amine ≥96.0%

Supplier: TCI America

T1724-25G T1724-5G
TCT1724-25GEA 611.62 CAD
TCT1724-25G TCT1724-005G
Tris(4-iodophenyl)amine ≥96.0%
Tris(4-iodophenyl)amine

CAS Number: 4181-20-8
MDL Number: MFCD01321198
Molecular Formula: C18H12I3N
Molecular Weight: 623.01
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: Very Pale Yellow

Formula: C₁₈H₁₂I₃N
MDL Number: MFCD01321198
CAS Number: 4181-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a6ebe65a9b0f3d51a7697a537ff5e3c0e66ef9d&vid=7a6ebe65a9b0f3d51a7697a537ff5e3c0e66ef9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR