(S)-(+)-3-Hydroxytetrahydrofuran ≥98.0%

Supplier: TCI America

T1716-5G T1716-1G
TCT1716-5GEA 62.88 CAD
TCT1716-5G TCT1716-001G
(S)-(+)-3-Hydroxytetrahydrofuran ≥98.0%
(S)-(+)-3-Hydroxytetrahydrofuran

CAS Number: 86087-23-2
MDL Number: MFCD00064327
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 80
Flash Point (°C): 79
Specific Gravity (20/20): 1.11
Specific rotation [a]20/D: 18 deg (C=2, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-Tetrahydrofuran-3-ol

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 80 °C
Density: 1.11 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00064327
CAS Number: 86087-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20db8c868be2fc0d57e52010ad522d607ddad7bb&vid=20db8c868be2fc0d57e52010ad522d607ddad7bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR