2,4,6-Triamino-5-nitrosopyrimidine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
T1705-5G
T1705-25G
TCT1705-5GEA
26.86
CAD
TCT1705-5G
TCT1705-25G
2,4,6-Triamino-5-nitrosopyrimidine ≥98.0% (by HPLC, titration analysis)
2,4,6-Triamino-5-nitrosopyrimidine
CAS Number: 1006-23-1
MDL Number: MFCD00006096
Molecular Formula: C4H6N6O
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Pale Red
Formula:
C₄H₆N₆O MW: 154.13 g/mol |
MDL Number:
MFCD00006096 CAS Number: 1006-23-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...