(2R,3R)-Tartranilic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
T1702-1G
T1702-5G
TCT1702-001GEA
124.82
CAD
TCT1702-001G
TCT1702-005G
(2R,3R)-Tartranilic acid ≥95.0% (by titrimetric analysis)
(2R,3R)-Tartranilic acid
[for Optical Resolution]
CAS Number: 3019-58-7
MDL Number: MFCD01321193
Molecular Formula: C10H11NO5
Molecular Weight: 225.20
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: 116 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks
Other Product Names / Synonyms: (2R,3R)-2,3-Dihydroxy-3-(phenylcarbamoyl)propionic Acid
Formula:
C₁₀H₁₁NO₅ |
MDL Number:
MFCD01321193 CAS Number: 3019-58-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...