Bis(2,4,6-trichlorophenyl)oxalate ≥98.0%

Supplier: TCI America

Synonyms: Bis(2,4,6-trichlorophenyl) oxalate, TCPO, Oxalic Acid Bis(2,4,6-trichlorophenyl) Ester

A5302-1G A5302-5G
TCA5302-001GEA 107.44 CAD
TCA5302-001G TCA5302-005G
Bis(2,4,6-trichlorophenyl)oxalate ≥98.0%
Bis(2,4,6-trichlorophenyl)oxalate

[Chemiluminescence reagent for the determination of fluorescent compounds]
CAS Number: 1165-91-9
MDL Number: MFCD00043061
Molecular Formula: C14H4Cl6O4
Molecular Weight: 448.88
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 190


Product Categories: Analytical Science, Oxalates (Chemiluminescence Reagents)
Other Product Names / Synonyms: Oxalic Acid Bis(2,4,6-trichlorophenyl) Ester, TCPO

Formula: C₁₄H₄Cl₆O₄
MW: 448.88 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00043061
CAS Number: 1165-91-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93c3dc845fd1787fd7e7e40b66743245cc394b00&vid=93c3dc845fd1787fd7e7e40b66743245cc394b00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR