1,4,8,11-Tetraazacyclopentadecane ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1,4,8,12-Tetraazacyclopentadecane
T1691-1G
T1691-5G
TCT1691-001GEA
505.6
CAD
TCT1691-001G
TCT1691-5G
1,4,8,11-Tetraazacyclopentadecane ≥97.0% (by titrimetric analysis)
1,4,8,11-Tetraazacyclopentadecane
CAS Number: 15439-16-4
MDL Number: MFCD00010049
Molecular Formula: C11H26N4
Molecular Weight: 214.36
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 101
Formula:
C₁₁H₂₆N₄ MW: 214.35 g/mol Melting Pt: 101 °C |
MDL Number:
MFCD00010049 CAS Number: 15439-16-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...