3-Bromo-2,2-bis(bromomethyl)propanol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1687-25G T1687-500G
TCT1687-025GEA 72.68 CAD
TCT1687-025G TCT1687-500G
3-Bromo-2,2-bis(bromomethyl)propanol ≥98.0% (by titrimetric analysis)
3-Bromo-2,2-bis(bromomethyl)propanol

CAS Number: 1522-92-5
MDL Number: MFCD00021982
Molecular Formula: C5H9Br3O
Molecular Weight: 324.84
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 131
Melting point (°C): 67


Other Product Names / Synonyms: 3-Bromo-2,2-bis(bromomethyl)-1-propanol, Tribromoneopentyl Alcohol, 2,2,2-Tris(bromomethyl)ethanol

Formula: C5H9Br3O
Boiling Pt: 131 °C
Melting Pt: 67 °C
MDL Number: MFCD00021982
CAS Number: 1522-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26754cba8718447f85c90f2ef44542d2ca024578&vid=26754cba8718447f85c90f2ef44542d2ca024578&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR