Tris(dibenzoylmethanato)iron ≥97.0%

Supplier: TCI America

Synonyms: Tris(1,3-diphenyl-1,3-propanedionato) Iron

T1686-25G T1686-5G
TCT1686-25GEA 694.25 CAD
TCT1686-25G TCT1686-005G
Tris(dibenzoylmethanato)iron ≥97.0%
Tris(dibenzoylmethanato)iron

CAS Number: 14405-49-3
MDL Number: MFCD00192557
Molecular Formula: C45H33FeO6
Molecular Weight: 725.60
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Deep Yellow Red


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Tris(1,3-diphenyl-1,3-propanedionato) Iron

Formula: C₄₅H₃₃FeO₆
MDL Number: MFCD00192557
CAS Number: 14405-49-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ca0a70fe6d231629d7891adc80264a84cac2853&vid=8ca0a70fe6d231629d7891adc80264a84cac2853&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR