Isoluminol (4-aminophthalhydrazide) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A5300-1G A5300-10G
TCA5300-001GEA 158 CAD
TCA5300-001G TCA5300-010G
Isoluminol (4-aminophthalhydrazide) ≥98.0% (by HPLC, titration analysis)
Isoluminol (4-Aminophthalhydrazide)

[Chemiluminescence reagent]
CAS Number: 3682-14-2
MDL Number: MFCD00010560
Molecular Formula: C8H7N3O2
Molecular Weight: 177.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 300


Product Categories: Analytical Science, Luminols (Chemiluminescence Reagents)
Other Product Names / Synonyms: 6-Amino-2,3-dihydro-1,4-phthalazinedione, 4-Aminophthalhydrazide, 4-Aminophthaloylhydrazine

Formula: C₈H₇N₃O₂
Melting Pt: 300 °C
MDL Number: MFCD00010560
CAS Number: 3682-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cf760ad7b9d721d9644b0d195f603ec20919c48&vid=2e5dc857054b80f013d2d899fb37ff5b80b03365&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR