2,2'-Thiobis(4-tert-octylphenol) ≥95.0%

Supplier: TCI America

T1674-25G T1674-500G
TCT1674-025GEA 59.57 CAD
TCT1674-025G TCT1674-500G
2,2'-Thiobis(4-tert-octylphenol) ≥95.0%
2,2'-Thiobis(4-tert-octylphenol)

CAS Number: 3294-03-9
MDL Number: MFCD00190577
Molecular Formula: C28H42O2S
Molecular Weight: 442.70
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 137


Other Product Names / Synonyms: Bis(2-hydroxy-5-tert-octylphenyl) Sulfide

Formula: C₂₈H₄₂O₂S
Melting Pt: 137 °C
MDL Number: MFCD00190577
CAS Number: 3294-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=772a3b6824cf59417ee95f4cc97636d72ac94d1e&vid=772a3b6824cf59417ee95f4cc97636d72ac94d1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR