5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene ≥95.0%

Supplier: TCI America

Synonyms: Trimethyl(1,2,3,4,5-pentamethylcyclopenta-2,4-dien-1-yl)silane, Trimethylsilylpentamethylcyclopentadiene

T1670-5ML T1670-1ML
TCT1670-005MLEA 44.24 CAD
TCT1670-005ML TCT1670-001ML
5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene ≥95.0%
5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene

CAS Number: 87778-95-8
MDL Number: MFCD00151193
Molecular Formula: C13H24Si
Molecular Weight: 208.42
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 101
Flash Point (°C): 76
Specific Gravity (20/20): 0.87
Storage Temperature: 0-10°C

Formula: C₁₃H₂₄Si
MW: 208.42 g/mol
Boiling Pt: 101 °C
Density: 0.87 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00151193
CAS Number: 87778-95-8

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR