Trimethylene bis(4-aminobenzoate) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
T1646-25G
T1646-100G
TCT1646-025GEA
75.84
CAD
TCT1646-025G
TCT1646-100G
Trimethylene bis(4-aminobenzoate) ≥98.0% (by HPLC, titration analysis)
Trimethylene bis(4-aminobenzoate)
CAS Number: 57609-64-0
MDL Number: MFCD00129765
Molecular Formula: C17H18N2O4
Molecular Weight: 314.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 128
Other Product Names / Synonyms: 1,3-Bis(4-aminobenzoyloxy)propane, 1,3-Propanediol Bis(4-aminobenzoate)
Formula:
C₁₇H₁₈N₂O₄ Melting Pt: 128 °C |
MDL Number:
MFCD00129765 CAS Number: 57609-64-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...