2-Acetylnaphthalene ≥98.0%

Supplier: TCI America

Synonyms: 2´-Acetonaphthone, 1-(Naphthalen-2-yl)ethanone, Methyl β-naphthyl ketone, 2-Acetonaphthone, 2'-Acetonaphthone

A0053-500G A0053-25G A0053-100G
TCA0053-500GEA 319.16 CAD
TCA0053-500G TCA0053-025G TCA0053-100G
2-Acetylnaphthalene ≥98.0%
2-Acetylnaphthalene

CAS Number: 93-08-3
MDL Number: MFCD00004108
Molecular Formula: C12H10O
Molecular Weight: 170.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 301
Melting point (°C): 54
Flash Point (°C): 129


Other Product Names / Synonyms: 2-Acetylnaphthalene, Methyl 2-Naphthyl Ketone

Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 301 °C
Melting Pt: 54 °C
Density: 1.1171 g/cm³ (20 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00004108
CAS Number: 93-08-3
EINECS: 202-216-2
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46c74c291cf0ed116133b04459c41366880ece6c&vid=46c74c291cf0ed116133b04459c41366880ece6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR