Triphenylsulfonium bromide ≥98.0%

Supplier: TCI America

T1609-5G T1609-1G T1609-100MG
TCT1609-5GEA 1606.86 CAD
TCT1609-5G TCT1609-1G TCT1609-100MG
Triphenylsulfonium bromide ≥98.0%
Triphenylsulfonium bromide

CAS Number: 3353-89-7
MDL Number: MFCD00428762
Molecular Formula: C18H15BrS
Molecular Weight: 343.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 293

Formula: C₁₈H₁₅BrS
MW: 343.28 g/mol
Melting Pt: 293 °C
MDL Number: MFCD00428762
CAS Number: 3353-89-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02e3fb2b66c97975e9737c9bd420cc7e9034e1a6&vid=02e3fb2b66c97975e9737c9bd420cc7e9034e1a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR