4-Hydroxy-6-(trifluoromethyl)pyrimidine ≥98.0%
Supplier: TCI America
T1601-5G
T1601-25G
TCT1601-5GEA
53.72
CAD
TCT1601-5G
TCT1601-25G
4-Hydroxy-6-(trifluoromethyl)pyrimidine ≥98.0%
4-Hydroxy-6-(trifluoromethyl)pyrimidine
CAS Number: 1546-78-7
MDL Number: MFCD00192530
Molecular Formula: C5H3F3N2O
Molecular Weight: 164.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 170
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Hydroxy-6-trifluoromethylpyrimidine
Formula:
C₅H₃F₃N₂O Melting Pt: 170 °C |
MDL Number:
MFCD00192530 CAS Number: 1546-78-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...