(S)-2-Methylbutyl-p-toluenesulfonate ≥95.0%
Supplier: TCI America
T1595-25G
T1595-5G
TCT1595-025GEA
323.9
CAD
TCT1595-025G
76220-502
(S)-2-Methylbutyl-p-toluenesulfonate ≥95.0%
(S)-2-Methylbutyl-p-toluenesulfonate
CAS Number: 38261-81-3
MDL Number: MFCD00013399
Molecular Formula: C12H18O3S
Molecular Weight: 242.33
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.11
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: p-Toluenesulfonic Acid (S)-2-Methylbutyl Ester
Formula:
C₁₂H₁₈O₃S Density: 1.11 (20 °C) |
MDL Number:
MFCD00013399 CAS Number: 38261-81-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...