Tetraethyl-p-xylylenediphosphonate ≥95.0%

Supplier: TCI America

Synonyms: p-Xylylenediphosphonic Acid Tetraethyl Ester, p-Bis(diethylphosphono)xylene

T1582-100G T1582-5G T1582-25G
TCT1582-100GEA 676.24 CAD
TCT1582-100G TCT1582-5G TCT1582-025G
Tetraethyl-p-xylylenediphosphonate ≥95.0%
Tetraethyl-p-xylylenediphosphonate

CAS Number: 4546-04-7
MDL Number: MFCD00671576
Molecular Formula: C16H28O6P2
Molecular Weight: 378.34
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: White
Boiling point (°C): 220
Melting point (°C): 76


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: p-Bis(diethylphosphono)xylene, p-Xylylenediphosphonic Acid Tetraethyl Ester

Formula: C₁₆H₂₈O₆P₂
Boiling Pt: 220 °C
Melting Pt: 76 °C
MDL Number: MFCD00671576
CAS Number: 4546-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d46405b14f58c28c88f37d53d924fb481d5eccc3&vid=d46405b14f58c28c88f37d53d924fb481d5eccc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR