2-Aminoperimidine hydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A5133-25G A5133-5G
TCA5133-25GEA 682.56 CAD
TCA5133-25G TCA5133-005G
2-Aminoperimidine hydrobromide ≥98.0% (by titrimetric analysis)
2-Aminoperimidine hydrobromide

[Precipitation reagent for SO4]
CAS Number: 40835-96-9
MDL Number: MFCD00012749
Molecular Formula: C11H9BrN3
Molecular Weight: 264.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 300

Formula: C₁₁H₉BrN₃
Melting Pt: 300 °C
MDL Number: MFCD00012749
CAS Number: 40835-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c13f9cc6f8fa8029a2353d1c0d9800f954a5519&vid=5c13f9cc6f8fa8029a2353d1c0d9800f954a5519&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR